Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
setd1ba
Entrez Gene ID
567970
Gene Name
SET domain containing 1B, histone lysine methyltransferase a
Gene Aliases
im:7156157, si:dkey-237o15.4
Chromosome Location
Chr.10: 134967 - 171347 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045134.2 NP_001038599.2 18 - 19 5497 66
XM_005169219.4 XP_005169276.1 17 - 18 5626 66
XM_005169220.4 XP_005169277.1 18 - 19 5218 66
XM_009305666.3 XP_009303941.1 17 - 18 5629 66
XM_009305667.3 XP_009303942.1 17 - 18 5444 66
XM_009305669.3 XP_009303944.1 18 - 19 4864 66

View Full Product Details