Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
setd1ba
Entrez Gene ID
567970
Gene Name
SET domain containing 1B, histone lysine methyltransferase a
Gene Aliases
im:7156157, si:dkey-237o15.4
Chromosome Location
Chr.10: 134967 - 171347 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045134.2 NP_001038599.2 1 - 2 207 86

View Full Product Details