Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
setd1ba
Entrez Gene ID
567970
Gene Name
SET domain containing 1B, histone lysine methyltransferase a
Gene Aliases
im:7156157, si:dkey-237o15.4
Chromosome Location
Chr.10: 134967 - 171347 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045134.2 NP_001038599.2 7 - 8 1937 81

View Full Product Details