Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kctd2
Entrez Gene ID
751724
Gene Name
potassium channel tetramerization domain containing 2
Gene Aliases
zgc:153278
Chromosome Location
Chr.12: 41107 - 62978 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045434.1 NP_001038899.1 2 - 3 716 78
GenBank mRNA BC122218.1 - 2 - 3 716 78

View Full Product Details