Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rwdd2b
Entrez Gene ID
566444
Gene Name
RWD domain containing 2B
Gene Aliases
si:dkey-177p2.14, zgc:153162, zgc:56075
Chromosome Location
Chr.20: 26905180 - 26916187 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075115.1 NP_001068583.1 2 - 3 316 70
GenBank mRNA BC124264.1 - 2 - 3 316 70

View Full Product Details