Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdgfab
Entrez Gene ID
767790
Gene Name
platelet-derived growth factor alpha polypeptide b
Gene Aliases
wu:fc32c12, zgc:153249
Chromosome Location
Chr.3: 102511 - 146600 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076757.1 NP_001070225.1 5 - 6 1110 97
GenBank mRNA BC124288.1 - 5 - 6 1110 97

View Full Product Details