Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
taar10a
Entrez Gene ID
794359
Gene Name
trace amine-associated receptor 10a
Gene Aliases
si:dkey-7c18.9
Chromosome Location
Chr.20: 46157434 - 46159150 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082898.1 NP_001076367.1 1 - 1 47 81
XM_017352532.2 XP_017208021.1 1 - 1 210 81

Gene Symbol
LOC100535889
Entrez Gene ID
100535889
Gene Name
trace amine-associated receptor 1-like
Gene Aliases
-
Chromosome Location
Chr.Unknown: 6061 - 7792 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_009303508.3 XP_009301783.1 1 - 1 214 81

View Full Product Details