Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ftr02
Entrez Gene ID
570286
Gene Name
finTRIM family, member 2
Gene Aliases
zgc:136797
Chromosome Location
Chr.2: 42236057 - 42242771 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003197921.5 XP_003197969.1 4 - 5 1167 82
GenBank mRNA AM941330.1 - 4 - 5 872 82
BC115273.1 - 4 - 5 1106 82

View Full Product Details