Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
zp2l2
Entrez Gene ID
559251
Gene Name
zona pellucida glycoprotein 2, like 2
Gene Aliases
zgc:162961, zp2v2, zpb2c
Chromosome Location
Chr.6: 9945317 - 9952103 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089502.1 NP_001082971.1 - 1242 91
GenBank mRNA BC134168.1 - - 1242 91

View Full Product Details