Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ankrd1b
Entrez Gene ID
564159
Gene Name
ankyrin repeat domain 1b (cardiac muscle)
Gene Aliases
ankrd1, zgc:171642
Chromosome Location
Chr.12: 16440744 - 16446373 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102388.1 NP_001095858.1 8 - 9 802 96
GenBank mRNA BC150367.1 - 8 - 9 802 96

View Full Product Details