Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
siglec15l
Entrez Gene ID
100124651
Gene Name
sialic acid binding Ig-like lectin 15, like
Gene Aliases
siglec-2
Chromosome Location
Chr.16: 41505263 - 41515698 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102649.1 NP_001096119.1 6 - 7 1076 63
GenBank mRNA DQ471942.1 - 6 - 7 1076 63

View Full Product Details