Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
es1
Entrez Gene ID
30237
Gene Name
es1 protein
Gene Aliases
C21orf22, esi, fk61d03, gatd3a, wu:fk61d03
Chromosome Location
Chr.8: 4838192 - 4850806 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131039.1 NP_571114.1 5 - 6 576 73
GenBank mRNA BC122433.1 - 5 - 6 557 73
U10403.1 - 5 - 6 576 73

View Full Product Details