Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mab21l1
Entrez Gene ID
246091
Gene Name
mab-21-like 1
Gene Aliases
-
Chromosome Location
Chr.15: 33070939 - 33073200 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152974.2 NP_694506.2 1 - 1 1344 74
GenBank mRNA AY048747.1 - 1 - 1 973 74
BC057249.1 - 1 - 1 1344 74
BC066667.1 - 1 - 1 1332 74

View Full Product Details