Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gnaia
Entrez Gene ID
323509
Gene Name
guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide a
Gene Aliases
cb874, fc01b07, wu:fc01b07
Chromosome Location
Chr.25: 20997671 - 21031044 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198805.2 NP_942100.1 1 - 2 473 59
GenBank mRNA BC049537.1 - 1 - 2 436 59
EH537458.1 - 1 - 2 64 59

View Full Product Details