The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199703.1 | NP_955997.1 | 9 - 10 | 1061 | 84 |
XM_005163387.4 | XP_005163444.1 | 9 - 10 | 1092 | 84 | |
XM_005163388.4 | XP_005163445.1 | 9 - 10 | 1092 | 84 | |
XM_005163389.4 | XP_005163446.1 | 9 - 10 | 1092 | 84 | |
XM_005163390.4 | XP_005163447.1 | 9 - 10 | 1092 | 84 | |
XM_005163391.4 | XP_005163448.1 | 9 - 10 | 1092 | 84 | |
XM_005163392.4 | XP_005163449.1 | 9 - 10 | 1092 | 84 | |
GenBank mRNA | BC055134.1 | - | 9 - 10 | 1061 | 84 |