Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ppdpfb
- Entrez Gene ID
-
336303
- Gene Name
- pancreatic progenitor cell differentiation and proliferation factor b
- Gene Aliases
-
c20orf149b, c20orf149l, wu:fj62f01, zgc:73395
- Chromosome Location
-
Chr.23: 17427838 - 17429740 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200008.1 |
NP_956302.1 |
3 - 4 |
299 |
82 |
GenBank mRNA |
BC059699.1 |
- |
3 - 4 |
299 |
82 |
View Full Product Details