Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppdpfb
Entrez Gene ID
336303
Gene Name
pancreatic progenitor cell differentiation and proliferation factor b
Gene Aliases
c20orf149b, c20orf149l, wu:fj62f01, zgc:73395
Chromosome Location
Chr.23: 17427838 - 17429740 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200008.1 NP_956302.1 4 - 5 398 69
GenBank mRNA BC059699.1 - 4 - 5 398 69

View Full Product Details