Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fam91a1
Entrez Gene ID
393413
Gene Name
family with sequence similarity 91 member A1
Gene Aliases
zgc:63609
Chromosome Location
Chr.19: 236853 - 281178 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200441.1 NP_956735.1 11 - 12 1146 63
XM_021468013.1 XP_021323688.1 11 - 12 1312 63
GenBank mRNA AY423008.1 - 10 - 11 853 63
BC054922.1 - 11 - 12 1146 63

View Full Product Details