Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mtap
Entrez Gene ID
393526
Gene Name
methylthioadenosine phosphorylase
Gene Aliases
zgc:66012
Chromosome Location
Chr.1: 26111024 - 26127939 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200554.1 NP_956848.1 6 - 7 723 88
GenBank mRNA BC046035.1 - 6 - 7 690 88
BC056545.1 - 6 - 7 723 88
BC164800.1 - 6 - 7 712 88

View Full Product Details