Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clic1
Entrez Gene ID
324481
Gene Name
chloride intracellular channel 1
Gene Aliases
wu:fc30e04, zgc:77044
Chromosome Location
Chr.19: 18758266 - 18765909 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212682.1 NP_997847.1 2 - 3 427 132
GenBank mRNA BC066618.1 - 2 - 3 427 132

View Full Product Details