Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nf2b
Entrez Gene ID
405887
Gene Name
neurofibromin 2b (merlin)
Gene Aliases
NF2a, nf2
Chromosome Location
Chr.21: 25801345 - 25816390 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212951.1 NP_998116.1 10 - 11 1110 116
GenBank mRNA AY561520.1 - 10 - 11 1110 116
BC079503.1 - 10 - 11 1047 116
BC108005.1 - 10 - 11 1116 116

View Full Product Details