Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nrp2b
Entrez Gene ID
405902
Gene Name
neuropilin 2b
Gene Aliases
id:ibd2759, npn-2B, sb:cb513
Chromosome Location
Chr.9: 14367578 - 14504971 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212966.1 NP_998131.1 11 - 12 2449 58
XM_017357663.2 XP_017213152.1 11 - 12 2578 58
GenBank mRNA AY351913.1 - 11 - 12 1946 58
AY428802.1 - 11 - 12 2449 58
AY437404.1 - 11 - 12 2409 58
BC163576.1 - 11 - 12 2264 58
BC163589.1 - 11 - 12 2264 58
KM655620.1 - 11 - 12 1859 58

View Full Product Details