Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc25a25a
Entrez Gene ID
406541
Gene Name
solute carrier family 25 member 25a
Gene Aliases
slc25a, slc25a25, wu:fb13d12, wu:fd14e03, zgc:77454
Chromosome Location
Chr.8: 2606067 - 2616128 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213257.1 NP_998422.1 6 - 7 868 77
GenBank mRNA BC066404.1 - 6 - 7 868 77

View Full Product Details