Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prkab1b
Entrez Gene ID
335232
Gene Name
protein kinase, AMP-activated, beta 1 non-catalytic subunit, b
Gene Aliases
prkab1, wu:fk93d05, wu:fw87e09, zgc:56652, zgc:76975
Chromosome Location
Chr.8: 39674707 - 39703747 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328554.1 NP_001315483.1 4 - 5 527 110
NM_214713.2 NP_999878.1 4 - 5 572 110
GenBank mRNA BC066758.1 - 4 - 5 487 110
EH456078.1 - 4 - 5 520 110

View Full Product Details