Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pkmb
Entrez Gene ID
445094
Gene Name
pyruvate kinase M1/2b
Gene Aliases
pkm2b, zgc:92037
Chromosome Location
Chr.25: 29159817 - 29196072 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003488.1 NP_001003488.1 2 - 3 269 70
GenBank mRNA BC076497.1 - 2 - 3 269 70
BC165710.1 - 2 - 3 279 70

View Full Product Details