Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ing4
Entrez Gene ID
322113
Gene Name
inhibitor of growth family, member 4
Gene Aliases
wu:fb11d09, wu:fb50e02, zgc:110619
Chromosome Location
Chr.19: 9512982 - 9522548 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020468.2 NP_001018304.1 1 - 2 65 76
GenBank mRNA BC095338.1 - 1 - 2 61 76
BC165444.1 - 1 - 2 71 76
EH590713.1 - 1 - 2 75 76

View Full Product Details