Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrpl27
Entrez Gene ID
553709
Gene Name
mitochondrial ribosomal protein L27
Gene Aliases
zgc:112322
Chromosome Location
Chr.12: 21684291 - 21688087 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020680.2 NP_001018516.1 1 - 2 41 68
GenBank mRNA BC095759.1 - 1 - 2 43 68

View Full Product Details