Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
psma3
Entrez Gene ID
564370
Gene Name
proteasome 20S subunit alpha 3
Gene Aliases
im:6909944, zgc:114044
Chromosome Location
Chr.17: 11421712 - 11439788 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030187.1 NP_001025358.1 6 - 7 580 74
GenBank mRNA BC097109.1 - 6 - 7 580 74

View Full Product Details