Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ptpn4b
Entrez Gene ID
407634
Gene Name
protein tyrosine phosphatase non-receptor type 4b
Gene Aliases
fi40g11, ptpn4a, si:rp71-1n18.2, wu:fi40g11
Chromosome Location
Chr.22: 11779834 - 11847366 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044313.2 NP_001037778.2 19 - 20 1991 95
XM_005173946.4 XP_005174003.1 20 - 21 2387 95
XM_009296096.3 XP_009294371.1 20 - 21 2384 95
GenBank mRNA FN428700.1 - 1 - 2 91 95

View Full Product Details