Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
setdb1a
Entrez Gene ID
553292
Gene Name
SET domain bifurcated histone lysine methyltransferase 1a
Gene Aliases
SETDB1, fc07d03, si:ch211-81a5.6, wu:fc07d03
Chromosome Location
Chr.19: 9111548 - 9144392 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044767.2 NP_001038232.1 16 - 17 4065 145

View Full Product Details