The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045109.2 | NP_001038574.2 | 3 - 4 | 822 | 93 |
GenBank mRNA | BC163243.1 | - | 3 - 4 | 532 | 93 |
DQ499477.1 | - | 3 - 4 | 782 | 93 |