Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nrxn2a
Entrez Gene ID
558326
Gene Name
neurexin 2a
Gene Aliases
-
Chromosome Location
Chr.21: 27635482 - 28341215 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079970.1 NP_001073439.1 20 - 21 4415 78
XM_005161353.4 XP_005161410.1 3 - 4 1884 78
XM_009295476.3 XP_009293751.1 20 - 21 4616 78
XM_009295477.3 XP_009293752.1 19 - 20 4592 78
XM_009295478.3 XP_009293753.1 19 - 20 4598 78
XM_009295479.3 XP_009293754.1 3 - 4 1879 78
XM_021468849.1 XP_021324524.1 20 - 21 4640 78
XM_021468850.1 XP_021324525.1 20 - 21 4406 78
XM_021468851.1 XP_021324526.1 19 - 20 4613 78
XM_021468852.1 XP_021324527.1 18 - 19 3693 78
XM_021468853.1 XP_021324528.1 18 - 19 4571 78
XM_021468855.1 XP_021324530.1 20 - 21 4646 78
GenBank mRNA DQ641428.1 - 20 - 21 4415 78
DQ641429.1 - 3 - 4 1818 78

View Full Product Details