Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
csf1b
Entrez Gene ID
790931
Gene Name
colony stimulating factor 1b (macrophage)
Gene Aliases
csf1-2, zgc:158436
Chromosome Location
Chr.8: 37168533 - 37202417 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080076.1 NP_001073545.1 2 - 3 366 122
XM_005167226.4 XP_005167283.1 2 - 3 430 122
GenBank mRNA AM901599.1 - 2 - 3 397 122
BC128871.1 - 2 - 3 366 122
KM655634.1 - 2 - 3 152 122

View Full Product Details