The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082809.1 | NP_001076278.1 | 5 - 6 | 461 | 74 |
GenBank mRNA | BC133925.1 | - | 5 - 6 | 414 | 74 |