Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ell2
Entrez Gene ID
322997
Gene Name
elongation factor for RNA polymerase II 2
Gene Aliases
cb244, fb78f03, im:3814960, sb:cb244, wu:fb78f03
Chromosome Location
Chr.21: 11749104 - 11774965 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083812.2 NP_001077281.2 2 - 3 239 65
XM_021468754.1 XP_021324429.1 2 - 3 211 65
GenBank mRNA BC056829.1 - 2 - 3 214 65
BC085600.1 - 2 - 3 216 65
BC108016.1 - 2 - 3 254 65
BC116571.1 - 2 - 3 244 65
BC124149.1 - 2 - 3 240 65
BC135070.1 - 2 - 3 214 65
BM096368.1 - 2 - 3 242 65

View Full Product Details