Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ell2
Entrez Gene ID
322997
Gene Name
elongation factor for RNA polymerase II 2
Gene Aliases
cb244, fb78f03, im:3814960, sb:cb244, wu:fb78f03
Chromosome Location
Chr.21: 11749104 - 11774965 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083812.2 NP_001077281.2 6 - 7 908 78
XM_021468754.1 XP_021324429.1 6 - 7 880 78
GenBank mRNA BC056829.1 - 6 - 7 883 78
BC085600.1 - 6 - 7 885 78
BC108016.1 - 6 - 7 923 78
BC116571.1 - 6 - 7 913 78
BC124149.1 - 6 - 7 909 78
BC135070.1 - 6 - 7 882 78
CT677237.2 - 2 - 3 275 78

View Full Product Details