Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
comtb
Entrez Gene ID
565370
Gene Name
catechol-O-methyltransferase b
Gene Aliases
si:dkey-13a21.15, zK13A21.15, zgc:162236
Chromosome Location
Chr.11: 234444 - 246172 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083843.1 NP_001077312.1 2 - 3 396 65
GenBank mRNA BC134931.1 - 2 - 3 396 65

View Full Product Details