Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rsph3
Entrez Gene ID
562259
Gene Name
radial spoke head 3
Gene Aliases
rshl2, si:ch211-122h15.2
Chromosome Location
Chr.20: 13632526 - 13640598 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089395.2 NP_001082864.2 8 - 9 1114 129
GenBank mRNA BC134867.1 - 8 - 9 1114 129

View Full Product Details