Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rhobtb4
Entrez Gene ID
553415
Gene Name
Rho related BTB domain containing 4
Gene Aliases
rhobtb2a
Chromosome Location
Chr.5: 43427891 - 43509101 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099974.2 NP_001093444.1 4 - 5 1304 114
NM_001365558.1 NP_001352487.1 4 - 5 950 114
GenBank mRNA AY865573.1 - 3 - 4 484 114

View Full Product Details