Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sp2
Entrez Gene ID
556835
Gene Name
sp2 transcription factor
Gene Aliases
-
Chromosome Location
Chr.11: 92088 - 116466 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099982.2 NP_001093452.2 4 - 5 1084 92
XM_009305862.1 XP_009304137.1 4 - 5 2062 92
GenBank mRNA EF613286.1 - 4 - 5 1078 92

View Full Product Details