Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
flr
Entrez Gene ID
797829
Gene Name
fleer
Gene Aliases
ttc30, ttc30a
Chromosome Location
Chr.3: 28831470 - 28850898 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104649.2 NP_001098119.2 11 - 12 1163 120
XM_021471707.1 XP_021327382.1 10 - 11 1136 120
XM_021471709.1 XP_021327384.1 9 - 10 926 120
XM_021471716.1 XP_021327391.1 11 - 12 1225 120
GenBank mRNA BC163912.1 - 11 - 12 1185 120
EF653429.1 - 11 - 12 1245 120
EU124003.1 - 11 - 12 1177 120

View Full Product Details