Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
flr
Entrez Gene ID
797829
Gene Name
fleer
Gene Aliases
ttc30, ttc30a
Chromosome Location
Chr.3: 28831470 - 28850898 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104649.2 NP_001098119.2 17 - 18 1784 82
XM_021471707.1 XP_021327382.1 17 - 18 1805 82
XM_021471709.1 XP_021327384.1 16 - 17 1595 82
XM_021471716.1 XP_021327391.1 18 - 19 1894 82
GenBank mRNA BC163912.1 - 17 - 18 1806 82
EF653429.1 - 17 - 18 1866 82

View Full Product Details