Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
flr
Entrez Gene ID
797829
Gene Name
fleer
Gene Aliases
ttc30, ttc30a
Chromosome Location
Chr.3: 28831470 - 28850898 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104649.2 NP_001098119.2 2 - 3 213 80
XM_021471716.1 XP_021327391.1 2 - 3 275 80
GenBank mRNA BC163912.1 - 2 - 3 235 80
EU124003.1 - 2 - 3 227 80

View Full Product Details