Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp8b2
Entrez Gene ID
100004274
Gene Name
cytochrome P450, family 8, subfamily B, polypeptide 2
Gene Aliases
zgc:174342
Chromosome Location
Chr.2: 3452590 - 3454556 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110288.1 NP_001103758.1 1 - 1 354 79
GenBank mRNA BC154281.1 - 1 - 1 354 79

View Full Product Details