Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbx16
Entrez Gene ID
30264
Gene Name
T-box transcription factor 16
Gene Aliases
cb103, id:ibd2185, spt, spt-1, wu:fa17g07, zgc:109817
Chromosome Location
Chr.8: 51736296 - 51753588 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131058.1 NP_571133.1 2 - 3 334 69
GenBank mRNA AF044977.1 - 2 - 3 249 69
AF077225.1 - 2 - 3 334 69
BC095003.1 - 2 - 3 350 69
BC165213.1 - 1 - 2 202 69

View Full Product Details