Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hoxa2b
Entrez Gene ID
30325
Gene Name
homeobox A2b
Gene Aliases
Z-75, hoxa2
Chromosome Location
Chr.16: 20926673 - 20929067 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131106.1 NP_571181.1 1 - 2 811 63
GenBank mRNA AF307010.1 - 1 - 2 811 63
BC162323.1 - 1 - 2 650 63
BC162328.1 - 1 - 2 648 63
DQ060536.1 - 1 - 2 228 63

View Full Product Details