Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ctbp1
Entrez Gene ID
65229
Gene Name
C-terminal binding protein 1
Gene Aliases
wu:fi43a11
Chromosome Location
Chr.14: 25084 - 49762 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131714.2 NP_571789.2 3 - 4 266 68
GenBank mRNA AB032415.1 - 3 - 4 68
BC045280.1 - 3 - 4 266 68

View Full Product Details