Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dixdc1a
Entrez Gene ID
360137
Gene Name
DIX domain containing 1a
Gene Aliases
ccd1, dixdc1
Chromosome Location
Chr.15: 18130109 - 18155985 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182884.1 NP_878304.1 4 - 5 847 115
GenBank mRNA AF502143.1 - 4 - 5 847 115

View Full Product Details