Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdcd4b
Entrez Gene ID
321061
Gene Name
programmed cell death 4b
Gene Aliases
cb45, pdcd4, sb:cb45, wu:fc84b08
Chromosome Location
Chr.22: 29658983 - 29689999 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198978.1 NP_945329.1 4 - 5 604 74
XM_005161791.4 XP_005161848.1 5 - 6 687 74
GenBank mRNA BC045513.1 - 4 - 5 604 74
BC065341.1 - 5 - 6 629 74

Gene Symbol
LOC103909982
Entrez Gene ID
103909982
Gene Name
programmed cell death protein 4-like
Gene Aliases
-
Chromosome Location
Chr.Unknown: 1709 - 6180 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_009298358.3 XP_009296633.1 2 - 3 74

View Full Product Details