Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eif5
Entrez Gene ID
322256
Gene Name
eukaryotic translation initiation factor 5
Gene Aliases
fb37c12, fb54h04, wu:fb37c12, wu:fb54h04, zgc:56606, zgc:77026
Chromosome Location
Chr.20: 18702734 - 18713949 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199591.2 NP_955885.1 2 - 3 495 92
XM_005160454.3 XP_005160511.1 3 - 4 619 92
GenBank mRNA AY577010.1 - 3 - 4 562 92
BC049502.1 - 1 - 2 134 92
BC066614.1 - 2 - 3 495 92

View Full Product Details